Microbiology detection tests, Bactident®
Bactident® Coagulase, 6
Supplier: Merck
Jump to:
Product Details & Documents
The presence or absence of enzyme activity is a taxonomic property of microorganisms and can be used for their differentiation or identification.
Documents
Certificates
Search by Lot NumberMERC1.13306.0001_TDS_EN
Datablad
VWR Non IvD Culture Media
Produktlitteratur
Specifications
Specifications
- Pk:6
- Description:Bactident® Coagulase