Order Entry
Norway
Orders LinkContactUsLinkComponent
Microbiology detection tests, Bactident®
Microbiology detection tests, Bactident®
Bactident® E.Coli, 50 Tests
Catalog # 1.13303.0001
Supplier:   Merck
Microbiology detection tests, Bactident®
Catalog # 1.13303.0001
Supplier:  Merck
CAS Number:  
Microbiology detection tests, Bactident®
Microbiology detection tests, Bactident®
Bactident® E.Coli, 50 Tests
CAS Number:  
Price
kr 2 570,00
Log in for å se tilgjengelighet og deres pris
1 * 1 Each
Product Details & Documents
The presence or absence of enzyme activity is a taxonomic property of microorganisms and can be used for their differentiation or identification.
Spesifikasjoner
Spesifikasjoner
  • Pk:
    50 Tests
  • Description:
    Bactident® E.Coli
Spesifikasjoner