Microbiology detection tests, Bactident®
Bactident® Aminopeptidase, 50 Tests
: Merck
The presence or absence of enzyme activity is a taxonomic property of microorganisms and can be used for their differentiation or identification.
Documents
- Pk:50 Tests
- Description:Bactident® Aminopeptidase