(±)-2-Octanol 97%
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol
- CAS Number:123-96-6
- Formula:C₈H₁₈O
- MDL Number:MFCD00004591
- Molecular Weight:130.23 g/mol
- Merck Index:13,06785
- Boiling Pt:174…181 °C (760 mmHg)
- Melting Pt:–38 °C
- Density:0.81 g/cm³
- Flash Pt:72 °C
- Storage Temperature:Ambient
- EINECS:204-667-0
- ADR:6.1,III
Documents
ACRO12941
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4250 to 1.4270 (20°C, 589 nm)
- GC:≥96.0 %
- Titration with NaOH:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Cat. No.:ACRO129410010



