Order Entry
Norway
ContactUsLinkComponent
IL-12 Total Mouse, Biotrak Assay, ELISA - 96 wells (Range: 47-3000pg/ml. Sensitivity: <12pg/ml. Sample volume: 50ul. 1.5L wash buff er supplied. Suitable for use with cell culture supernatant, serum and plasma (EDTA, heparin and sodium citrate)) 1 * 96 Assays
Catalog # RPN2702
Supplier:  Cytiva
IL-12 Total Mouse, Biotrak Assay, ELISA - 96 wells (Range: 47-3000pg/ml. Sensitivity: <12pg/ml. Sample volume: 50ul. 1.5L wash buff er supplied. Suitable for use with cell culture supernatant, serum and plasma (EDTA, heparin and sodium citrate)) 1 * 96 Assays
Catalog # RPN2702
Supplier:  Cytiva