(±)-Linalool 97%
Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
- CAS Number:78-70-6
- Formula:C₁₀H₁₈O
- MDL Number:MFCD00008906
- Molecular Weight:154.25 g/mol
- Merck Index:13,05517
- Boiling Pt:199 °C (760 mmHg)
- Density:0.861 g/cm³
- Flash Pt:75 °C
- Storage Temperature:Ambient
- EINECS:201-134-4
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4600 to 1.4640 (20°C, 589 nm)
- GC:≥96.0 %
- Specific gravity:(20/4°C) 0.859 to 0.864
- Specific optical rotation:-2° to +2° (20°C, 589 nm) (neat)
- Substance darkened by H2SO4:≤10 APHA
- Calcium (Ca):≤0.5 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.02 ppm
- Magnesium (Mg):≤0.05 ppm
- Potassium (K):≤0.1 ppm
- Sodium (Na):≤0.5 ppm
- Zinc (Zn):≤0.2 ppm
- Total phosphorus (P):≤0.1 ppm
- Total silicon (Si):≤0.05 ppm
- Total sulfur (S):≤0.001 %
- Bromine number:≤0.5
Specifications
- Pack type:Glass bottle
- Pk:500 mL



