Order Entry
Norway
ContactUsLinkComponent
IL-8, Human, Biotrak Assay, ELISA - 96 wells (Range: 26-1000pg/ml. Sensitivity: <5pg/ml. Sample volume: 50ul. 1.5L wash buffer supp lied. Suitable for use with cell culture supernatant, serum, plasma and urine) 1 * 1 items
Catalog # RPN2764
Supplier:  Cytiva
IL-8, Human, Biotrak Assay, ELISA - 96 wells (Range: 26-1000pg/ml. Sensitivity: <5pg/ml. Sample volume: 50ul. 1.5L wash buffer supp lied. Suitable for use with cell culture supernatant, serum, plasma and urine) 1 * 1 items
Catalog # RPN2764
Supplier:  Cytiva