4'-Bromoacetophenone 98%
Glass bottle for solids, 25 g
: p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
- :99-90-1
- :C₈H₇BrO
- :MFCD00000105
- :199.05 g/mol
- :13,01387
- :255 °C (760 mmHg)
- :48…52 °C
- :1,736 g/cm³ (20 °C)
- :>100 °C
- :Ambient
- :202-799-3
- :6.1,III
- Appearance:White to light yellow Crystals or flakes
- Infrared spectrum:Conforms
- Melting point:48 to 52 °C
- GC:≥97.5 %
- Pack type:Glass bottle for solids
- Pk:25 g



