Order Entry
Norway
Orders LinkContactUsLinkComponent
Spring clamps
Stainless steel spring clamp, 10 ml Erlenmeyer, round bottomed flasks or beakers
  444-9491
Spring clamps
  444-9491
Spring clamps
Spring clamps
Stainless steel spring clamp, 10 ml Erlenmeyer, round bottomed flasks or beakers
kr 215,00
1 * 1 Items
  • For:
    10 ml Erlenmeyer, round bottomed flasks or beakers
  • Description:
    Stainless steel spring clamp