Order Entry
Norway
Orders LinkContactUsLinkComponent
D-(-)-Mannitol ACS

D-(-)-Mannitol ACS

Plastic bottle, 250 g
Catalog # ACRO423922500
D-(-)-Mannitol ACS
Catalog # ACRO423922500
CAS Number:  69-65-8
D-(-)-Mannitol ACS
D-(-)-Mannitol ACS

Plastic bottle, 250 g

CAS Number:  69-65-8
Price
kr 800,00
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290 - 295 °C (4 hPa)
  • Melting Pt:165…170 °C
  • Density:1,49 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-711-8

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.05 % (5 g, 105°C, 2 hrs)
  • Residue after ignition:
    ≤0.01 %
  • Specific optical rotation:
    +23.3° to +24.3° (25°C, 589 nm)
  • Heavy metals (ICP-OES):
    ≤5 ppm (as Pb)
  • Titrable acid:
    ≤0.0008 meq/g
  • Insoluble matter:
    ≤0.01 %
  • Reducing sugars:
    Passes test

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.