Lead(II) nitrate ACS
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Lead(II) nitrate ACS
Catalog # ACRO423850050
Supplier: Thermo Fisher Scientific
CAS Number: 10099-74-8
Jump to:
Product Details & Documents
Synonyms: Lead dinitrate
- CAS Number:10099-74-8
- Formula:Pb(NO₃)₂
- MDL Number:MFCD00011153
- Molecular Weight:331.21 g/mol
- Merck Index:13,05432
- Melting Pt:470 °C (dec.)
- Density:4.53 g/cm³
- Storage Temperature:Ambient
- EINECS:233-245-9
- UN:1469
- ADR:5.1,II
Documents
ACRO42385
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Titration Complexometric:≥99.0 %
- Chloride (Cl):≤10 ppm
- Calcium (Ca):≤50 ppm
- Copper (Cu):≤20 ppm
- Iron (Fe):≤10 ppm
- Potassium (K):≤50 ppm
- Sodium (Na):≤200 ppm
- Insoluble matter:≤0.005 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



