DL-α-Tocopherol acetate 98%
Glass bottle, 1 g
Flash Point: >110°C
Appearance: Clear light yellow to light green Viscous liquid
: (±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate
- :7695-91-2
- :C₃₁H₅₂O₃
- :MFCD00072042
- :472.75 g/mol
- :257 °C
- :Ambient
- Appearance:Clear light yellow to light green Viscous liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- HPLC:≥97.5 %
- Pack type:Glass bottle
- Pk:1 g



