Order Entry
Norway
Orders LinkContactUsLinkComponent
Inoculating loops and needles

Inoculating loops and needles

Loop, Ø 1,5 mm
Catalog # 631-1401
Supplier:   BOCHEM
Inoculating loops and needles
Catalog # 631-1401
Supplier:  BOCHEM
Inoculating loops and needles
Inoculating loops and needles

Loop, Ø 1,5 mm

Price
kr 29,00
Log in for å se tilgjengelighet og deres pris
1 * 1 Items
Product Details & Documents

Stainless steel 18/10. Length 100 mm.

Spesifikasjoner
  • Description:
    Loop, Ø 1,5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.