Diphenyl ditelluride 97%
Glass bottle for solids, 1 g
- :32294-60-3
- :C₁₂H₁₀Te₂
- :MFCD00192106
- :409.41 g/mol
- :64…67 °C
- :Ambient
- :250-982-1
- :3284
- :6.1,III
Documents
ACRO30184
SDS
- Appearance:Orange to brown Powder or crystals
- Infrared spectrum:Conforms
- Melting point:64 to 67 °C
- HPLC:≥96.0 %
- Pack type:Glass bottle for solids
- Pk:1 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



