Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PH-ELEKTRODE TIL PHSCAN 3 OG 3PLUS 1 * 1 Items
Catalog # EUTEEC-PHWPSEN03
Supplier:
EUTECH
Expand image
PH-ELEKTRODE TIL PHSCAN 3 OG 3PLUS 1 * 1 Items
Catalog # EUTEEC-PHWPSEN03
Supplier:
EUTECH