2-Hydroxypropionic acid 85%
Bottle
Catalog # APOSOR1024496-500G
New Product
Supplier: Apollo Scientific
2-Hydroxypropionic acid 85%
Catalog # APOSOR1024496-500G
Supplier: Apollo Scientific
New Product
CAS Number: 50-21-5
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid
- CAS Number:50-21-5
- Formula:C₃H₆O₃
- Molecular Weight:90,08 g/mol
Specifications
Specifications
- Pack type:Bottle



