Order Entry
Norway
Orders LinkContactUsLinkComponent
2-Hydroxypropionic acid 85%

2-Hydroxypropionic acid 85%

Bottle
Catalog # APOSOR1024496-100G
New Product
Supplier:   Apollo Scientific
2-Hydroxypropionic acid 85%
Catalog # APOSOR1024496-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  50-21-5
2-Hydroxypropionic acid 85%

Bottle

CAS Number:  50-21-5
Price
kr 270,00
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid
  • CAS Number:50-21-5
  • Formula:C₃H₆O₃
  • Molecular Weight:90,08 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications