VCMMAE ≥98%
Bottle
Catalog # APOSBUP05884-500MG
New Product
Supplier: Apollo Scientific
VCMMAE ≥98%
Catalog # APOSBUP05884-500MG
Supplier: Apollo Scientific
New Product
CAS Number: 646502-53-6
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms: VcMMAE
- CAS Number:646502-53-6
- Formula:C₆₈H₁₀₅N₁₁O₁₅
- Molecular Weight:1316,63 g/mol
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



