VCMMAE ≥98%
Bottle
Catalog # APOSBUP05884-100MG
New Product
Supplier: Apollo Scientific
VCMMAE ≥98%
Catalog # APOSBUP05884-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 646502-53-6
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms: VcMMAE
- CAS Number:646502-53-6
- Formula:C₆₈H₁₀₅N₁₁O₁₅
- Molecular Weight:1316,63 g/mol
Specifications
Specifications
- Pack type:Bottle



