Order Entry
Norway
Orders LinkContactUsLinkComponent
Paracetamol 98%

Paracetamol 98%

Bottle
Catalog # APOSOR3962-100G
Irritant
New Product
Supplier:   Apollo Scientific
Paracetamol 98%
Catalog # APOSOR3962-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  103-90-2
Paracetamol 98%

Bottle

CAS Number:  103-90-2
Price
kr 305,00
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP
  • CAS Number:103-90-2
  • Formula:C₈H₉NO₂
  • MDL Number:MFCD00002328
  • Molecular Weight:151.16 g/mol
  • Melting Pt:168…172 °C
  • Density:1.293 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:203-157-5
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.