Order Entry
Norway
Orders LinkContactUsLinkComponent
Paracetamol 98%

Paracetamol 98%

Bottle
Catalog # APOSOR3962-25G
Irritant
New Product
Supplier:   Apollo Scientific
Paracetamol 98%
Catalog # APOSOR3962-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  103-90-2
Paracetamol 98%

Bottle

CAS Number:  103-90-2
Price
kr 270,00
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP
  • CAS Number:103-90-2
  • Formula:C₈H₉NO₂
  • MDL Number:MFCD00002328
  • Molecular Weight:151.16 g/mol
  • Melting Pt:168…172 °C
  • Density:1,293 g/cm³ (25 °C)
  • Storage Temperature:Romtemperatur
  • EINECS:203-157-5
Specifications

Specifications

  • Pakketype:
    Bottle

Specifications