Boc-Pro-OH
Bottle
APOSOR61282-25KG
New Product
: N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid
- :15761-39-4
- :C₁₀H₁₇NO₄
- :MFCD00037324
- :215.25 g/mol
- :133…136 °C
- :Refrigerator
- :239-848-3
- Pack type:Bottle