Order Entry
Norway
Orders LinkContactUsLinkComponent
D-(-)-N-Methylglucamine

D-(-)-N-Methylglucamine

Bottle
Catalog # APOSOR5855-100G
Irritant
New Product
Supplier:   Apollo Scientific
D-(-)-N-Methylglucamine
Catalog # APOSOR5855-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  6284-40-8
D-(-)-N-Methylglucamine

Bottle

CAS Number:  6284-40-8
Price
kr 450,00
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol
  • CAS Number:6284-40-8
  • Formula:H₃CNHCH₂(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004707
  • Molecular Weight:195.22 g/mol
  • Melting Pt:129…131.5 °C
  • Storage Temperature:Ambient
  • EINECS:228-506-9
Specifications

Specifications

  • Pack type:
    Bottle

Specifications