Order Entry
Norway
Orders LinkContactUsLinkComponent
 

2-Hydroxypropionic acid 85%

New Product
2-Hydroxypropionic acid 85%
New Product
 
2-Hydroxypropionic acid 85%

 

 

Usage: Organic building block/reagent

 :  DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid
  •  :50-21-5
  •  :C₃H₆O₃
  •  :90,08 g/mol
Product Family Options

 

  • Pack type
     
     
  • Bottle
       
       
    1 * 100 g
    1 * 100 g - kr270.00
    kr270.00
    1 * 100 g
       kr270.00

     
    New Product
    New Product
  • Bottle
       
       
    1 * 500 g
    1 * 500 g - kr450.00
    kr450.00
    1 * 500 g
       kr450.00

     
    New Product
    New Product