Order Entry
Norway
Orders LinkContactUsLinkComponent
Linalool ≥98%

Linalool ≥98%

Bottle
Catalog # APOSBUP19001-1G
Irritant
New Product
Supplier:   Apollo Scientific
Linalool ≥98%
Catalog # APOSBUP19001-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  78-70-6
Linalool ≥98%

Bottle

CAS Number:  78-70-6
Price
kr 470,00
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
  • CAS Number:78-70-6
  • Formula:C₁₀H₁₈O
  • Molecular Weight:154,25 g/mol
  • EINECS:201-134-4
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.