Order Entry
Norway
Orders LinkContactUsLinkComponent
 

ADA ≥98%

Bottle
Catalog # APOSBUP18857-500MG
New Product
Supplier:   Apollo Scientific
ADA ≥98%
Catalog # APOSBUP18857-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  26239-55-4
ADA ≥98%

Bottle

CAS Number:  26239-55-4
Price
kr 470,00
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid
  • CAS Number:26239-55-4
  • Formula:C₆H₁₀N₂O₅
  • Molecular Weight:190,15 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.