Order Entry
Norway
Orders LinkContactUsLinkComponent
 

Linalool 98%

Irritant
New Product
Supplier:   Apollo Scientific
Linalool 98%
Supplier:  Apollo Scientific
New Product
Certificates

3 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
  • CAS Number:78-70-6
  • Formula:C₁₀H₁₈O
  • Molecular Weight:154,25 g/mol
  • EINECS:201-134-4
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - kr 270,00
    kr 270,00
    1 * 25 mL
       kr 270,00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - kr 305,00
    kr 305,00
    1 * 100 mL
       kr 305,00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 450,00
    kr 450,00
    1 * 500 mL
       kr 450,00

    Specifications:
    New Product
    New Product