Order Entry
Norway
ContactUsLinkComponent
VWR® DOSATEST® Test Strips, Cobalt
VWR® DOSATEST® Test Strips, Cobalt
Catalog # 92539.601
Supplier:  VWR Chemicals
New Product
VWR® DOSATEST® Test Strips, Cobalt
Catalog # 92539.601
Supplier:  VWR Chemicals
New Product

Specifications

  • Description:
    Single parameter cobalt semi-quantitative test strips, 100 tests

Specifications

About this item

Semi-quantitative test strips for the determination of Cobalt.

  • Range: Cobalt - 0, 10, 25, 50, 100, 250, 500 and 1000  mg/L Co²⁺

    Shelf life: 2.5 Year(s)

    Storage temperature: 4 to 30 °C (39 to 86 °F)

    Size: 100 test strips in a tube, instruction leaflet

Ideal for rapid and easy analysis of Cobalt directly on-site. No maintenance or calibration is required.