Scissors, Medical Grade, Extra Fine Blades
Catalog # EXCE361
Supplier: EXCELTA
New Product
Specifications
- Length:81,25 mm
- Blade length:25,4 mm
Specifications
About this item
These medical grade scissors are made of stainless steel.
Excellent for ultra-miniature work in confined areas
- Cleanroom safe, medical grade stainless steel
-
Extremely precise cutting edges
Scissors, medical grade, 4 star quality
17° curved
Stainless steel AISI 420
They are ideal for ultra miniature work in confined areas.