Specifications
- Colour:Blue
- Description:Nucleolus Blue™ DCS1, cell-impermeant, optimal for dead or fixed cells
- Storage conditions:Freeze (<−15 °C); minimize light exposure
- Application:Fluorescence microscopy, flow cytometry, high-content imaging
Specifications
About this item
Nucleolus Blue™ DCS1 is a cell-impermeant fluorescent dye that selectively binds to RNA within the nucleolus, a non-membrane-bound nuclear compartment responsible for ribosomal RNA (rRNA) transcription and processing. Upon binding to RNA, the dye exhibits strong blue fluorescence, whereas its interaction with DNA results in only weak fluorescence, providing exceptional specificity for nucleolar RNA detection.
Selective nucleolar RNA staining - binds nucleolar RNA with strong blue fluorescence while exhibiting minimal DNA fluorescence
Optimised for fixed and permeabilised cells - ideal for dead cell staining and fixed-cell imaging
High-contrast nucleolar visualisation - enables precise detection of nucleoli, facilitating research on nucleolar organization, function, and morphology
Compatible with multi-colour staining - can be used in combination with nuclear stains such as Nuclear Green™ LCS1 or Nuclear Red™ LCS1 for enhanced cellular context
This dye is particularly useful for studying nucleolar morphology and function in dead or permeabilised cells. It enables clear visualisation of nucleoli and can be used in conjunction with nuclear counterstains such as Nuclear Green™ LCS1 or Nuclear Red™ LCS1 to provide additional structural context.
The nucleolus plays a critical role in cellular processes, including ribosome biogenesis, DNA damage response, autophagy, viral pathogenesis, and cellular senescence. Morphological changes in the nucleolus have also been implicated as potential biomarkers for cancer diagnostics. Nucleolus Blue™ DCS1 is compatible with fluorescence microscopy, flow cytometry, and high-content imaging assays, making it a valuable tool for researchers investigating nucleolar dynamics and cellular homeostasis in various biological and disease contexts.