Order Entry
Norway
ContactUsLinkComponent
Ceramic Magnetic Hotplate Stirrer, AREC 7
Ceramic Magnetic Hotplate Stirrer, AREC 7
Catalog # VELPF20500680
Supplier:  VELP SCIENTIFIC
New Product
undefined
Ceramic Magnetic Hotplate Stirrer, AREC 7
Catalog # VELPF20500680
Supplier:  VELP SCIENTIFIC
New Product

Specifications

  • Control type:
    Analogue
  • Temperature range:
    Ambient...550 °C
  • Description:
    AREC 7 magnetic hotplate stirrer
  • W×D×H:
    160×270×100 mm
  • Plate dimensions:
    180×180 mm
  • Plate material:
    Ceramic
  • Power input/output:
    800 W
  • Weight:
    3,5 kg

Specifications

About this item

Analogue hotplate stirrer, equipped with a 177,8 mm square ceramic plate, safe and reliable, designed to deliver excellent heating and stirring performance for basic laboratory applications.

  • Advanced heating technology ensures temperature homogeneity and stability from room temperature up to 550 °C

  • Powerful brushless ensuring power and resistance over time
  • Stirs up to 15 L volume (H₂O) from 100 up to 1500 min⁻¹
  • SpeedServo™ counter reaction technology maintains the speed constant as viscosity changes
  • The robust 177,8 mm ceramic top offers exceptional chemical and scratch resistance and it is easy to clean with a simple wipe-down
  • A warning icon illuminates when the plate temperature exceeds 50 °C and the heating function is inactive

Optimum heating and stirring performance for basic laboratory applications. The AREC 7 ceramic hotplate stirrer is designed to deliver reliable performance for everyday laboratory applications, offering powerful heating and efficient stirring capabilities. Its advanced heating plate technology ensures uniform temperature control up to 550 °C, providing excellent heat stability for accurate sample preparation. Capable of stirring up to 15 L, the robust brushless motor operates smoothly across a wide speed range from 100 to 1500 min⁻¹. Whether performing titrations or preparing samples and buffers, the SpeedServo™ counter-reaction technology maintains consistent stirring speeds even when the medium’s viscosity changes, ensuring reliable and repeatable results every time.

Ease-of-use and safety for laboratory operators the AREC 7 ceramic hotplate stirrer prioritises operator safety and durability. A clear warning icon illuminates when the plate temperature exceeds 50 °C and the heating function is inactive, minimising the risk of accidental burns. The robust structure is designed to withstand harsh laboratory environments and resist chemical corrosion. The ceramic plate is scratch-proof and simple to clean, ensuring long-lasting performance with minimal maintenance.

Housing material: Aluminium body, technopolymer enclosure

Stirring volume: 15 L

Stirring speed range: 100 - 1500 min⁻¹

Motor: Brushless

Torque compensation: SpeedServo™

Hot top warning system: Illuminated icon (when the temperature is above 50 °C)