LN₂ Supply Tanks
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
Transport cryogenic industrial gases with Thermo Scientific™ LN₂ Supply tanks, which are designed to be both durable and user-friendly. For long-term storage of samples in cryogenic freezers, or controlling temperature during freezing procedures utilising controlled-rate freezing equipment.
Rated at 22 psi (151,7 kPa)
- Evaporate 2% of volume per day
Caution:
Warranty varies by country, please contact us for details.