Human Complement C1 ELISA Kit
ABCAAB222865-96
: Abcam
:
- Description:Human Complement C1 ELISA kit
- Tests per kit:96 tests
Human Complement C1 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Complement C1 in Human in Saliva, Urine, Plasma, Milk, Serum samples.