Human HBB ELISA Kit
Human HBB ELISA Kit, 96 tests
Catalog # ABCAAB235654-96
Supplier: Abcam
Jump to:
Product Details & Documents
Human HBB ELISA Kit is a single-wash 90-min Simplestep used to quantify Human HBB with a sensitivity of 34,1 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.
Specifications
Specifications
- Description:Human HBB ELISA Kit
- Tests per kit:96 tests
- Primary antibody reactivity:Human
- Sample type:Cell culture extracts, Heparin Plasma, Citrate plasma, Serum, EDTA Plasma
- Detection range:0,125 - 8 ng/ml
- Storage temperature:4 °C
- Assay Type:Sandwich
- Time to Results:1 h 30 min
- Cat. No.:ABCAAB235654-96
- Assay duration:One step
- Detection method:Colorimetric
- Sensitivity:34,1 pg/ml
- Format:Pre-coated
- Target protein:Hemoglobin
- Regulatory status:RUO



