Order Entry
Norway
ContactUsLinkComponent
Anti-IgG Donkey Polyclonal Antibody (Alexa Fluor® 555)
Anti-IgG Donkey Polyclonal Antibody (Alexa Fluor® 555)
Catalog # ABCAAB150070-500
Supplier:  Abcam
undefined
Anti-IgG Donkey Polyclonal Antibody (Alexa Fluor® 555)
Catalog # ABCAAB150070-500
Supplier:  Abcam

Specifications

  • Seq:
    1418
  • Size:
    500 µg
  • Pack type:
    Vial

Specifications

About this item

Donkey F(ab')2 Anti-Rabbit IgG - H&L (Alexa Fluor® 555), pre-adsorbed.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Rabbit