Gå til Overskrift
Gå til Hovedinnhold
Gå til Bunntekst
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human TGF beta Receptor II (from HEK293 cells)
Recombinant Human TGF beta Receptor II (from HEK293 cells)
Recombinant Human TGF beta Receptor II (from HEK293 cells)
Leverandør:
ANTIBODIES.COM
New Product
Sertifikat
Expand image
Recombinant Human TGF beta Receptor II (from HEK293 cells)
Leverandør:
ANTIBODIES.COM
New Product
Om dette produktet