1,2-Benzisoxazole ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:271-95-4
:
Formula:C₇H₅NO
MDL Number:MFCD00005852
:119.12 g/mol
Boiling Pt:100°C
Flash Pt:86°C
Storage Temperature:Refrigerator
EINECS:205-983-1
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %