Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Di-tert-butylfluorene
3,6-Di-tert-butylfluorene ≥98%
3,6-Di-tert-butylfluorene ≥98%
3,6-Di-tert-butylfluorene ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
3,6-Di-tert-butylfluorene ≥98%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
58775-07-8
:
Formula:
C₂₁H₂₆
MDL Number:
MFCD02683109
Melting Pt:
79…82 °C
:
278.44 g/mol
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAB23996
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation