Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Logg inn
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hjem
N-Nervonoyl taurine
N-Nervonoyl taurine
N-Nervonoyl taurine
N-Nervonoyl taurine
Leverandør:
Cayman Chemical
Sertifikat
Expand image
N-Nervonoyl taurine
Leverandør:
Cayman Chemical
Om dette produktet
:
Formel:
C₂₆H₅₁NO₄S