

Bis(2-ethylhexyl)phosphate ≥95.0% (by titrimetric analysis) for rare metals extraction
Supplier: TCI
Certificates
About this item
CAS Number:298-07-7
:
Formula:C₁₆H₃₅O₄P
MDL Number:MFCD00009492
:322.43 g/mol
Melting Pt:–50°C
Flash Pt:196°C
EINECS:206-056-4
UN:1902
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (Neutralization titration):min. 95.0 %
- Specific gravity (20/20):0.9730 to 0.9830
- Refractive index n20/D:1.4420 to 1.4440