

α-Phenylbenzyl bromide ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:776-74-9
:
Formula:(C₆H₅)₂CHBr
MDL Number:MFCD00000134
:247.13 g/mol
Boiling Pt:315 °C (1013 hPa)
Melting Pt:36…39 °C
Density:1,491 g/cm³ (13 °C)
Storage Temperature:Refrigerator
EINECS:212-279-8
UN:1770
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:White or Colorles to Yellow to Orange powder to lump to clear liquid
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %