Order Entry
Norway
ContactUsLinkComponent
Glycerine ≥99.0% (by GC) for electrophoresis
Glycerine ≥99.0% (by GC) for electrophoresis
Catalog # TCIAG0316-1G
Supplier:  TCI
CAS Number:  
undefined
Glycerine ≥99.0% (by GC) for electrophoresis
Catalog # TCIAG0316-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.0 %
  • Refractive index n20/D:
    1.4700 to 1.4750
  • Functionality test:
    to pass test

Specification Test Results

About this item

 :
 :92.09 g/mol
Merck Index:14,04484
Boiling Pt:290°C
Melting Pt:18.6 °C
Density:1,26 g/cm³ (20 °C)
Flash Pt:177°C
Storage Temperature:Ambient
EINECS:200-289-5
 :GHS07
CAS Number:56-81-5
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722