



1,4-Dimethylcyclohexane cis- and trans-mixture ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:589-90-2
:
Formula:C₈H₁₆
MDL Number:MFCD00001508
:112.22 g/mol
Boiling Pt:120 °C (1013 hPa)
Melting Pt:–87 °C
Density:0,77 g/cm³ (20 °C)
Flash Pt:10°C
EINECS:209-663-2
UN:2263
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomers)
Specification Test Results
Frequently Bought Together