2-Pentene cis- and trans-mixture ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:109-68-2
:
Formula:C₅H₁₀
MDL Number:MFCD00009384
:70.13 g/mol
Merck Index:14,07123
Boiling Pt:36°C
Melting Pt:–140 °C
Density:0,658 g/cm³ (20 °C)
Flash Pt:–45°C
Storage Temperature:Ambient
EINECS:203-695-0
UN:3295
ADR:3,II
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 % (total of isomers)