Order Entry
Norway
ContactUsLinkComponent
2,6,6-Trimethyl-2-cyclohexene-1,4-dione ≥98.0% (by GC)
Irritant
2,6,6-Trimethyl-2-cyclohexene-1,4-dione ≥98.0% (by GC)
Catalog # TCIAK0034-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2,6,6-Trimethyl-2-cyclohexene-1,4-dione ≥98.0% (by GC)
Catalog # TCIAK0034-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :152.19 g/mol
Boiling Pt:214 °C (1013 hPa)
Melting Pt:15 °C
Density:1,03 g/cm³ (20 °C)
EINECS:214-406-2
 :GHS07
CAS Number:1125-21-9
Formula:C₉H₁₂O₂
MDL Number:MFCD00043119