Order Entry
Norway
ContactUsLinkComponent
3-(Diethylamino)phenol ≥98.0% (by GC, titration analysis)
Irritant
Health
3-(Diethylamino)phenol ≥98.0% (by GC, titration analysis)
Catalog # TCIAD0470-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Health
3-(Diethylamino)phenol ≥98.0% (by GC, titration analysis)
Catalog # TCIAD0470-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to lump
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    72.0 to 75.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :165.24 g/mol
Boiling Pt:280°C
Melting Pt:78 °C
EINECS:202-090-9
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
CAS Number:91-68-9
Formula:C₁₀H₁₅NO
MDL Number:MFCD00002265