
cis-4-Hydroxycyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3685-22-1
:MFCD04038017
Formula:C₇H₁₂O₃
MDL Number:MFCD02093477
:144.1700 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:148.0 to 152.0 deg-C
- Solubility in Methanol:almost transparency