Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Amino-2-(methylthio)pyrimidine-5-carbonitrile
4-Amino-2-(methylthio)pyrimidine-5-carbonitrile
4-Amino-2-(methylthio)pyrimidine-5-carbonitrile
4-Amino-2-(methylthio)pyrimidine-5-carbonitrile
Supplier:
Thermo Scientific
Certificates
Expand image
4-Amino-2-(methylthio)pyrimidine-5-carbonitrile
Supplier:
Thermo Scientific
About this item
CAS Number:
770-30-9
:
Formula:
C₆H₆N₄S
MDL Number:
MFCD00023237
:
166.21 g/mol
UN:
3439
ADR:
6.1,III