Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D-tert-Leucine (H-D-Tle-OH)
D-tert-Leucine (H-D-Tle-OH)
D-tert-Leucine (H-D-Tle-OH)
D-tert-Leucine (H-D-Tle-OH)
Catalog # MOLE45088529-250MG
Supplier:
Molekula
CAS Number:
Expand image
D-tert-Leucine (H-D-Tle-OH)
Catalog # MOLE45088529-250MG
Supplier:
Molekula
CAS Number:
Specifications
Pk:
250 mg
Seq:
2590
Specifications
About this item
Product Details
:
:
131.2 g/mol
CAS Number:
26782-71-8
Formula:
C₆H₁₃NO₂
MDL Number:
MFCD00004265
Product Details