N-(Ethoxycarbonyl)phthalimide ≥98.0% (by total nitrogen basis) for peptide synthesis
Leverandør: TCI
Sertifikat
Om dette produktet
CAS-nummer:22509-74-6
:
Formel:C₁₁H₉NO₄
MDL-nummer:MFCD00005893
:219.20 g/mol
EINECS:245-048-5
Spesifikasjonstestresultater
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:82.0 to 92.0 deg-C
- Solubility in hot Methanol:almost transparency